Learn More
Apexbio Technology LLC ACETAMINOPHEN-D3-1MG
SDP
Catalog No. 5000569033
Encompass_Preferred
Acetaminophen-d3 (CAS No 60902-28-5) is a deuterated form of acetaminophen frequently used in biomedical research especially within pharmacokinetic and drug metabolism studies Originating as a derivative of the widely used analgesic and antipyretic compound its primary mechanism involves inhibition of the cyclooxygenase (COX) enzymes primarily within the central nervous system This deuterated variant is valuable for studying metabolic pathways due to its stability and distinct mass allowing for precise tracking via mass spectrometry It demonstrates activity ranging from nanomolar to low micromolar levels in vitro Acetaminophen-d3 is commonly used in cell models animal studies and high-throughput drug screening assays to elucidate metabolic processes and interactions with other pharmaceuticals Experimental concentrations typically depend on the specific research design
Catalog No. 50-005-69033 Supplier Apexbio Technology LLC Supplier No. SL10661
Acetaminophen-d3 (CAS No 60902-28-5) is a deuterated form of acetaminophen frequently used in biomedical research especially within pharmacokinetic and drug metabolism studies Originating as a derivative of the widely used analgesic and antipyretic compound its primary mechanism involves inhibition of the cyclooxygenase (COX) enzymes primarily within the central nervous system This deuterated variant is valuable for studying metabolic pathways due to its stability and distinct mass allowing for precise tracking via mass spectrometry It demonstrates activity ranging from nanomolar to low micromolar levels in vitro Acetaminophen-d3 is commonly used in cell models animal studies and high-throughput drug screening assays to elucidate metabolic processes and interactions with other pharmaceuticals Experimental concentrations typically depend on the specific research design