Learn More
Medchemexpress LLC Acetaminophen-d3 | 60902-28-5 | MFCD06658893 | 99.0% | 154.18 g/mol | C8H6D3NO2 | 10 MG
SDP
Catalog No. 5000298645
Encompass_Preferred
Acetaminophen-d3 is the deuterium-labeled analogue of acetaminophen used as a tracer and internal standard for quantitative analysis by NMR, GC-MS, and LC-MS. It is supplied as a high-purity solid suitable for analytical research applications.
  • High-purity deuterium-labeled standard (99.0%).
  • Intended for use as tracer or internal standard in NMR, GC-MS, and LC-MS quantitative analysis.
  • Supplied as a solid for analytical workflows and small-scale assays.
  • Stable when stored as a powder at -20°C for extended periods.
Catalog No. 50-002-98645 Supplier Medchemexpress LLC Supplier No. HY66005S110MG
Add to Cart
Add to Cart
Acetaminophen-d3 is the deuterium-labeled analogue of acetaminophen used as a tracer and internal standard for quantitative analysis by NMR, GC-MS, and LC-MS. It is supplied as a high-purity solid suitable for analytical research applications.
  • High-purity deuterium-labeled standard (99.0%).
  • Intended for use as tracer or internal standard in NMR, GC-MS, and LC-MS quantitative analysis.
  • Supplied as a solid for analytical workflows and small-scale assays.
  • Stable when stored as a powder at -20°C for extended periods.