Learn More
Cayman Chemical Acetaminophen-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 64315-36-2, 10 mg
SDP
Catalog No. 502979741
Encompass_Preferred
An internal standard for the quantification of acetaminophen by GC- or LC-MS
Catalog No. 50-297-9741 Supplier Cayman Chemical Supplier No. 1824510 MG
An internal standard for the quantification of acetaminophen by GC- or LC-MS