Learn More
Cayman Chemical Acetaminophen-d4, 64315-36-2, 5 mg
SDP
Supplier:  Cayman Chemical 182455
Encompass_Preferred
An internal standard for the quantification of acetaminophen by GC- or LC-MS
Catalog No. NC1471156