Learn More
Cayman Chemical Acetaminophen-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 64315-36-2, 5 mg
SDP
Catalog No. 502979893
Encompass_Preferred
An internal standard for the quantification of acetaminophen by GC- or LC-MS
Catalog No. 50-297-9893 Supplier Cayman Chemical Supplier No. 182455 MG
An internal standard for the quantification of acetaminophen by GC- or LC-MS