Learn More
Medchemexpress LLC Acetaminophen-d4 | 64315-36-2 | 99.60% | C8H5D4NO2 | 25 MG
SDP
Catalog No. 5000359261
Encompass_Preferred
Acetaminophen-d4 is the deuterium labeled Acetaminophen, a selective cyclooxygenase-2 (COX-2) inhibitor and a widely used antipyretic and analgesic agent. It is also a potent hepatic N-acetyltransferase 2 (NAT2) inhibitor. Deuteration, the incorporation of stable heavy isotopes into drug molecules, has gained attention for its potential to affect pharmacokinetic and metabolic profiles.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes into drug molecules for quantitation.
Catalog No. 50-003-59261 Supplier Medchemexpress LLC Supplier No. 25MGHY66005S
Acetaminophen-d4 is the deuterium labeled Acetaminophen, a selective cyclooxygenase-2 (COX-2) inhibitor and a widely used antipyretic and analgesic agent. It is also a potent hepatic N-acetyltransferase 2 (NAT2) inhibitor. Deuteration, the incorporation of stable heavy isotopes into drug molecules, has gained attention for its potential to affect pharmacokinetic and metabolic profiles.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes into drug molecules for quantitation.