Learn More
Medchemexpress LLC Acetaminophen-d7 | 1219798-53-4 | 99.9% | C8H2D7NO2 | 1 MG
SDP
Catalog No. 5000434727
Encompass_Preferred
Acetaminophen-d7 is the deuterium labeled Acetaminophen. Acetaminophen (Paracetamol) is a selective cyclooxygenase-2 (COX-2) inhibitor with an IC50 of 25.8 μM; it is a widely used antipyretic and analgesic agent and a potent hepatic N-acetyltransferase 2 (NAT2) inhibitor.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-34727 Supplier Medchemexpress LLC Supplier No. HY66005S21MG
Acetaminophen-d7 is the deuterium labeled Acetaminophen. Acetaminophen (Paracetamol) is a selective cyclooxygenase-2 (COX-2) inhibitor with an IC50 of 25.8 μM; it is a widely used antipyretic and analgesic agent and a potent hepatic N-acetyltransferase 2 (NAT2) inhibitor.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.