Learn More
Medchemexpress LLC Acetaminophen glucuronide | 16110-10-4 | MFCD03094680 | 99.9% | 327.29 g·mol-1 | C14H17NO8 | 5 MG
SDP
Catalog No. 5000262075
Encompass_Preferred
Acetaminophen glucuronide is the O-β-D-glucuronide metabolite of acetaminophen provided as a high-purity analytical reference standard (CAS 16110-10-4). It is supplied as a solid reference material for metabolite identification, pharmacokinetic studies, and analytical method development.
  • Reference standard for analytical and metabolic studies.
  • High purity (99.9%) suitable for quantitative analysis.
  • Solid form provided in analytical-scale pack sizes.
  • Used in pharmacokinetic and toxicology research.
  • Compatible with LC-MS and other analytical techniques.
Catalog No. 50-002-62075 Supplier Medchemexpress LLC Supplier No. HY1130835MG
Acetaminophen glucuronide is the O-β-D-glucuronide metabolite of acetaminophen provided as a high-purity analytical reference standard (CAS 16110-10-4). It is supplied as a solid reference material for metabolite identification, pharmacokinetic studies, and analytical method development.
  • Reference standard for analytical and metabolic studies.
  • High purity (99.9%) suitable for quantitative analysis.
  • Solid form provided in analytical-scale pack sizes.
  • Used in pharmacokinetic and toxicology research.
  • Compatible with LC-MS and other analytical techniques.