Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Acetaminophen glucuronide-d3 sodium salt | 1260619-61-1 | 97.8% | 352.29 g/mol | C14H13D3NNaO8 | 2.5 MG
SDP

Catalog No. 5000300587
Encompass_Preferred

Acetaminophen glucuronide-d3 sodium salt is the deuterium-labeled sodium salt of acetaminophen glucuronide, supplied as a stable isotope standard for analytical and metabolic research. It is intended as a reference material for quantitative mass spectrometry and chromatographic assays to improve accuracy and traceability in metabolite measurement.

  • Deuterium-labeled sodium salt suitable as an internal standard.
  • Enables accurate MS and LC quantitation of acetaminophen metabolites.
  • Molecular formula C14H13D3NNaO8; molecular weight 352.29 g/mol.
  • Provided in a research-grade, solid form for laboratory use.
  • For research use only; not for human or clinical use.

Catalog No. 50-003-00587 Supplier Medchemexpress LLC Supplier No. HYW016034S2.5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Acetaminophen glucuronide-d3 sodium salt is the deuterium-labeled sodium salt of acetaminophen glucuronide, supplied as a stable isotope standard for analytical and metabolic research. It is intended as a reference material for quantitative mass spectrometry and chromatographic assays to improve accuracy and traceability in metabolite measurement.

  • Deuterium-labeled sodium salt suitable as an internal standard.
  • Enables accurate MS and LC quantitation of acetaminophen metabolites.
  • Molecular formula C14H13D3NNaO8; molecular weight 352.29 g/mol.
  • Provided in a research-grade, solid form for laboratory use.
  • For research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.