Learn More
Medchemexpress LLC Acetaminophen glucuronide sodium salt | 120595-80-4 | MFCD00153898 | 98.2% | 349.27 | C14H16NNaO8 | 5 MG
SDP
Catalog No. 5000302882
Encompass_Preferred
Acetaminophen glucuronide sodium salt is the sodium salt of the major glucuronide metabolite of acetaminophen, supplied as a solid analytical reference for research use. It is intended for use in drug metabolism, pharmacokinetic, and toxicology studies, and as a standard for biological sample analysis when high-quality reference material is required.
  • White to off-white solid suitable for analytical use.
  • High purity for reference standard applications.
  • Available in small, quantified pack sizes for method development.
  • Store sealed at -20°C to maintain stability and prevent moisture uptake.
  • Useful as a metabolite standard in pharmacokinetic and toxicology studies.
Catalog No. 50-003-02882 Supplier Medchemexpress LLC Supplier No. HYW0160345MG
Acetaminophen glucuronide sodium salt is the sodium salt of the major glucuronide metabolite of acetaminophen, supplied as a solid analytical reference for research use. It is intended for use in drug metabolism, pharmacokinetic, and toxicology studies, and as a standard for biological sample analysis when high-quality reference material is required.
  • White to off-white solid suitable for analytical use.
  • High purity for reference standard applications.
  • Available in small, quantified pack sizes for method development.
  • Store sealed at -20°C to maintain stability and prevent moisture uptake.
  • Useful as a metabolite standard in pharmacokinetic and toxicology studies.