Learn More
Medchemexpress LLC Acetaminophen glucuronide sodium salt | 120595-80-4 | MFCD00153898 | ≥99.0% | 349.27 g/mol | C14H16NNaO8 | 1 MG
SDP
Catalog No. 5000302881
Encompass_Preferred
Acetaminophen glucuronide sodium salt is the sodium salt of the primary glucuronide metabolite of acetaminophen. Supplied as a white to off-white solid, it is used as a reference standard in analytical chemistry, metabolite identification, and toxicology research.
  • Primary glucuronide metabolite of acetaminophen.
  • Supplied as a white to off-white solid, soluble in water.
  • Molecular formula C14H16NNaO8 and molecular weight 349.27 g/mol.
  • High purity (≥99.0%).
  • Available in small pack sizes for analytical use (1 mg, 5 mg, 10 mg).
  • Suitable as an analytical reference, calibration standard, and for toxicology assays.
Catalog No. 50-003-02881 Supplier Medchemexpress LLC Supplier No. HYW0160341MG
Acetaminophen glucuronide sodium salt is the sodium salt of the primary glucuronide metabolite of acetaminophen. Supplied as a white to off-white solid, it is used as a reference standard in analytical chemistry, metabolite identification, and toxicology research.
  • Primary glucuronide metabolite of acetaminophen.
  • Supplied as a white to off-white solid, soluble in water.
  • Molecular formula C14H16NNaO8 and molecular weight 349.27 g/mol.
  • High purity (≥99.0%).
  • Available in small pack sizes for analytical use (1 mg, 5 mg, 10 mg).
  • Suitable as an analytical reference, calibration standard, and for toxicology assays.