Learn More
Cayman Chemical Acetaminophen, Purity: ≥98%, CAS: 103-90-2, 500 g
SDP
Catalog No. 502890557
Encompass_Preferred
An analgesic and antipyretic compound; an inhibitor of COX-2 that is selective over COX-1 (IC50s = 113.7 and 25.8 µM, respectively, in human blood ex vivo); induces ferroptotic cell death in primary mouse hepatocytes; has a metabolite that depletes glutathione reserves in the liver
Catalog No. 50-289-0557 Supplier Cayman Chemical Supplier No. 10024500 G
An analgesic and antipyretic compound; an inhibitor of COX-2 that is selective over COX-1 (IC50s = 113.7 and 25.8 µM, respectively, in human blood ex vivo); induces ferroptotic cell death in primary mouse hepatocytes; has a metabolite that depletes glutathione reserves in the liver