Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetaminophen, MP Biomedicals™

Catalog No. ICN19009190 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
ICN19009190 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19009190 Supplier MP Biomedicals, Inc Supplier No. 0219009190

missing translation for 'validMainframeMsg'

Specifications

CAS 103-90-2
Color White
Density 1.293g/cm3 at 21°C
Assay Percent Range 100.3%
Molecular Formula C8H9NO2
Quantity 500 g
MDL Number MFCD00002328
Synonym acetaminophen, 4-acetamidophenol, paracetamol, apap, n-4-hydroxyphenyl acetamide, tylenol, acetaminofen, panadol, datril, p-acetamidophenol
InChI Key RZVAJINKPMORJF-UHFFFAOYSA-N
SMILES CC(=O)NC1=CC=C(O)C=C1
IUPAC Name N-(4-hydroxyphenyl)acetamide
Molecular Weight (g/mol) 151.17
PubChem CID 1983
ChEBI CHEBI:46195
Formula Weight 151.2
Physical Form Powder
Chemical Name or Material Acetaminophen
Show More Show Less
Recommended Storage Store at 4°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.