Learn More
Medchemexpress LLC Acetaminophen (Standard) | 103-90-2 | 100.0% | C8H9NO2 | 25 MG
SDP
Catalog No. 5000359260
Encompass_Preferred
Acetaminophen (Standard) is an analytical standard of Acetaminophen, primarily used for research and analytical applications. Also known as Paracetamol or 4-Acetamidophenol, it is a selective cyclooxygenase-2 (COX-2) inhibitor and a widely recognized antipyretic and analgesic agent. It also acts as a potent hepatic N-acetyltransferase 2 (NAT2) inhibitor.
  • Analytical standard for research and analytical applications
  • Reference standard for assays
  • Suitable for HPLC, GC, and MS experiments
  • Functions as a selective cyclooxygenase-2 (COX-2) inhibitor
  • Widely used as an antipyretic and analgesic agent
  • Acts as a potent hepatic N-acetyltransferase 2 (NAT2) inhibitor
Catalog No. 50-003-59260 Supplier Medchemexpress LLC Supplier No. 25MGHY66005R
Acetaminophen (Standard) is an analytical standard of Acetaminophen, primarily used for research and analytical applications. Also known as Paracetamol or 4-Acetamidophenol, it is a selective cyclooxygenase-2 (COX-2) inhibitor and a widely recognized antipyretic and analgesic agent. It also acts as a potent hepatic N-acetyltransferase 2 (NAT2) inhibitor.
  • Analytical standard for research and analytical applications
  • Reference standard for assays
  • Suitable for HPLC, GC, and MS experiments
  • Functions as a selective cyclooxygenase-2 (COX-2) inhibitor
  • Widely used as an antipyretic and analgesic agent
  • Acts as a potent hepatic N-acetyltransferase 2 (NAT2) inhibitor