Learn More
Cayman Chemical Acetaminophen sulfate (potassium salt), 32113-41-0, 1 mg
SDP
Supplier:  Cayman Chemical 225711
Encompass_Preferred
Molecular Formula C8H8NO5S • K, Purity ≥98%, Formula Weight 269.3, Synonyms: N-[4-(sulfooxy)phenyl]-acetamide, monopotassium salt, 4-Acetamidophenol sulfate, APAP sulfate, 4'-Hydroxyacetanilide sulfate, Paracetamol sulfate; A metabolite of acetaminophen; excreted in urine.
Catalog No. NC1531266
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.