Learn More
Medchemexpress LLC HY-B0823 100mg Medchemexpress, Acetamiprid CAS:135410-20-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0823100MG
Encompass_Preferred
Medchemexpress, HY-B0823 100mg Acetamiprid CAS:135410-20-7 Acetamiprid is a neonicotinoid insecticide used worldwide. Acetamiprid is a nicotinic acetylcholine receptor (nAChR) agonist, and is shown to be associated with neuromuscular and reproductive disorders[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-1130