Learn More
Cayman Chemical Acetamiprid, Purity: ≥98%, CAS: 135410-20-7, 50 mg
SDP
Catalog No. 502934146
Encompass_Preferred
A neonicotinoid insecticide; an agonist of insect nAChRs; activates nAChRs containing N. lugens α1 and rat β2 subunits (EC50 = 67 µM in X. laevis oocytes); leads to sexual and aggressive behaviors in adult male offspring after administration to pregnant mice (1 mg/kg); decreases anxiety-like behavior of adult male offspring in the light-dark transition test (1 and 10 mg/kg)
Catalog No. 50-293-4146 Supplier Cayman Chemical Supplier No. 2412950 MG
A neonicotinoid insecticide; an agonist of insect nAChRs; activates nAChRs containing N. lugens α1 and rat β2 subunits (EC50 = 67 µM in X. laevis oocytes); leads to sexual and aggressive behaviors in adult male offspring after administration to pregnant mice (1 mg/kg); decreases anxiety-like behavior of adult male offspring in the light-dark transition test (1 and 10 mg/kg)