Learn More
Cayman Chemical AcetamIprId-d3 10mg
SDP
Catalog No. 502915928
Encompass_Preferred
An internal standard for the quantification of acetamiprid by GC- or LC-MS
Catalog No. 50-291-5928 Supplier Cayman Chemical Supplier No. 3956510 MG
An internal standard for the quantification of acetamiprid by GC- or LC-MS