Learn More
Medchemexpress LLC Acetamiprid-d3 | 1353869-35-8 | 99.9% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB0823S1MG
Encompass_Preferred
Acetamiprid-d3 is the deuterium labeled Acetamiprid. Acetamiprid is a neonicotinoid insecticide and a nAChR agonist.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration, the incorporation of stable heavy isotopes into drug molecules, has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-07651