Learn More
Cayman Chemical Acetamiprid-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 1353869-35-8, 1 mg
SDP
Catalog No. 502918280
Encompass_Preferred
An internal standard for the quantification of acetamiprid by GC- or LC-MS
Catalog No. 50-291-8280 Supplier Cayman Chemical Supplier No. 395651 MG
An internal standard for the quantification of acetamiprid by GC- or LC-MS