Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Acetanilide
SCI_ED

Catalog No. S25117 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Glistening Crystals
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25117 100 g Glistening Crystals
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S25117

missing translation for 'validMainframeMsg'

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 103-84-4
Molecular Formula C8H9NO
Molecular Weight (g/mol) 135.17
MDL Number MFCD00008674
InChI Key FZERHIULMFGESH-UHFFFAOYSA-N
Synonym acetanilide, acetanil, acetamidobenzene, antifebrin, acetylaniline, acetanilid, n-acetylaniline, acetamide, n-phenyl, acetic acid anilide, acetylaminobenzene
PubChem CID 904
ChEBI CHEBI:28884
IUPAC Name N-phenylacetamide
SMILES CC(=O)NC1=CC=CC=C1

Specifications

Color White
Density 1.21
Assay Percent Range 1
Packaging Glistening Crystals
Linear Formula CH3CONHC6H5
Quantity 100 g
Formula Weight 135.17
Chemical Name or Material Acetanilide
Safety handling Check Alert storage code Red
ShelfLife 5 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.