Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Acetanilide

Catalog No. 1129335G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-293-35G 5 g
11-293-3500G 500 g
11-293-31KG 1 kg
11-293-3100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 11-293-35G Supplier Sigma Aldrich Supplier No. 1129335G
May include imposed supplier surcharges.
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Acetanilide

Specifications

CAS 103-84-4
Melting Point 113°C to 115°C (lit.)
Boiling Point 304°C (lit.)
Molecular Formula C8H9NO
Linear Formula CH3CONHC6H5
MDL Number MFCD00008674
Quantity 5 g
Synonym N-Phenylacetamide
Molecular Weight (g/mol) 135.16
Percent Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.