Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetanilide, 99%, Spectrum™ Chemical
SDP

Catalog No. 18608316 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
250 g
500 g
1 kg
12 kg
50 kg
Packaging:
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-608-316 1 kg Poly Pail
18-608-318 250 g Poly Pail
18-608-320 500 g Poly Pail
18-608-317 12 kg Poly Pail
18-608-319 50 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-608-316 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. A10071KG
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 103-84-4

Specifications

CAS Max % 1
Melting Point 113° to 118°C
Boiling Point 304°C
Packaging Poly Pail
Quantity 1 kg
Percent Purity 0.99
Grade Reagent
Physical Form Crystalline Powder or Lumps

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.