Learn More
Perkin Elmer US LLC Acetanilide - Calibration Standard, 2 g
Catalog No. 502112001
Encompass_Preferred
The PerkinElmer Combustion Standards are stable materials which are not subject to ambient influences and are recommended for the PerkinElmer 2400/2410 and Model 240 instruments.
Catalog No. 50-211-2001 Supplier Perkin Elmer US LLC Supplier No. 02401121
The PerkinElmer Combustion Standards are stable materials which are not subject to ambient influences and are recommended for the PerkinElmer 2400/2410 and Model 240 instruments.