Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetanilide melting point standard Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111012353 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2353 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2353 Supplier MilliporeSigma Supelco Supplier No. PHR10861G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Acetanilide belongs to a class of compounds which are the active components of drugs exhibiting analgesic and antipyretic properties.

Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

Ignition Point 1004°F
CAS 103-84-4
Vapor Pressure 1 mmHg (114°C)
Vapor Density 4.65 (vs. air)
Flash Point 161°C - Closed Cup
Physical Form Neat
Linear Formula CH3 CONHC6H5
MDL Number MFCD00008674
Melting Point ∼115°C; 113°C to 115°C
Boiling Point 304°C
Synonym Acetanilide; N-Phenylacetamide
Type Certified Reference Material
Quantity 1 g
Formula Weight 135.16
Grade Certified Reference Material
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.