Learn More
U.S. Pharmacopeia Acetanilide Melting Point Standard, 103-84-4, MFCD00008674, 1 g
Catalog No. NC0741785
Encompass
Molecular Formula: C8 H9 N O, Molecular Weight: 135.165, mp: 113-115 °C, bp: 304 °C, Synonym: N-Phenylacetamide, Acetanilide.
Catalog No. NC0741785 Supplier U.S. Pharmacopeia Supplier No. 1004001
Add to Cart
Add to Cart
Molecular Formula: C8 H9 N O, Molecular Weight: 135.165, mp: 113-115 °C, bp: 304 °C, Synonym: N-Phenylacetamide, Acetanilide.