Learn More
Apexbio Technology LLC Acetarsone 97-44-9 1g
SDP
Catalog No. 5000096095
Encompass_Preferred
Acetarsone (97-44-9) is a small-molecule anti-infective and antiparasitic agent belonging to the organoarsenic group of compounds It is proposed to modulate parasite-specific metabolic enzymes or interfere with biochemical pathways essential for parasite survival Acetarsone exerts its biological activity primarily through inhibition of parasite biochemical pathways however the exact molecular targets and mechanism remain to be fully elucidated In both in vitro and in vivo studies acetarsone demonstrates antiparasitic activity with IC50 values reported in the low to moderate micromolar range depending on parasite species and experimental conditions Based on these pharmacological properties acetarsone holds research potential in the treatment of protozoan infections and in studies of intestinal inflammation and mucosal pathology
Catalog No. 50-000-96095 Supplier Apexbio Technology LLC Supplier No. A83551000
Acetarsone (97-44-9) is a small-molecule anti-infective and antiparasitic agent belonging to the organoarsenic group of compounds It is proposed to modulate parasite-specific metabolic enzymes or interfere with biochemical pathways essential for parasite survival Acetarsone exerts its biological activity primarily through inhibition of parasite biochemical pathways however the exact molecular targets and mechanism remain to be fully elucidated In both in vitro and in vivo studies acetarsone demonstrates antiparasitic activity with IC50 values reported in the low to moderate micromolar range depending on parasite species and experimental conditions Based on these pharmacological properties acetarsone holds research potential in the treatment of protozoan infections and in studies of intestinal inflammation and mucosal pathology