Learn More
Electron Microscopy Sciences Acetate Buffer, 0.2M, pH 3.6-5.6 225 ML
SDP
Catalog No. 50980215
Encompass_Preferred
Acetic Acid and Sodium Acetate, 0.2M, pH range 3.6-5.6. Our buffers are ideal for maintaining a constant pH during fixation and have an ionic constitution to prevent the extraction of cellular components or the deposition of precipitates of fixative chemicals. The buffers are prepared fresh for each order and conveniently packaged. Desired pH should be specified when ordering.
pH: 3.6-5.6
Catalog No. 50-980-215 Supplier Electron Microscopy Sciences Supplier No. 11480
Acetic Acid and Sodium Acetate, 0.2M, pH range 3.6-5.6. Our buffers are ideal for maintaining a constant pH during fixation and have an ionic constitution to prevent the extraction of cellular components or the deposition of precipitates of fixative chemicals. The buffers are prepared fresh for each order and conveniently packaged. Desired pH should be specified when ordering.
pH: 3.6-5.6