Learn More
Electron Microscopy Sciences Acetate Buffer, 0.2M, pH 3.6-5.6 500 ML
SDP
Catalog No. 50980216
Encompass_Preferred
Acetic Acid and Sodium Acetate, 0.2M, pH range 3.6-5.6. Our buffers are ideal for maintaining a constant pH during fixation and have an ionic constitution to prevent the extraction of cellular components or the deposition of precipitates of fixative chemicals. The buffers are prepared fresh for each order and conveniently packaged. Desired pH should be specified when ordering.
pH: 3.6-5.6
Catalog No. 50-980-216 Supplier Electron Microscopy Sciences Supplier No. 11482
Acetic Acid and Sodium Acetate, 0.2M, pH range 3.6-5.6. Our buffers are ideal for maintaining a constant pH during fixation and have an ionic constitution to prevent the extraction of cellular components or the deposition of precipitates of fixative chemicals. The buffers are prepared fresh for each order and conveniently packaged. Desired pH should be specified when ordering.
pH: 3.6-5.6