Learn More
Bioworld Acetate Buffer, 1M, pH 4.0, 1 L
Catalog No. 50255309
Encompass_Preferred
Product DescriptionAcetate buffer has a pH value of 4.0 and is made from a mixture of acetic acid (weak acid) and acetate ions (conjugated base). It is routinely used in DNA and RNA precipitation, and in the preparation of gel stains for protein gel electrophoresis. Experiments have shown that when added to food waste composting systems, sodium acetate may be effective at counteracting the adverse effects of organic acids produced in the composting process.It has a pKa value of 4.76 and a melting point of 323-329°C.For laboratory use only. Not fit for agricultural, household, clinical use or human consumption
Catalog No. 50-255-309 Supplier Bioworld Supplier No. 401200002
Product DescriptionAcetate buffer has a pH value of 4.0 and is made from a mixture of acetic acid (weak acid) and acetate ions (conjugated base). It is routinely used in DNA and RNA precipitation, and in the preparation of gel stains for protein gel electrophoresis. Experiments have shown that when added to food waste composting systems, sodium acetate may be effective at counteracting the adverse effects of organic acids produced in the composting process.It has a pKa value of 4.76 and a melting point of 323-329°C.For laboratory use only. Not fit for agricultural, household, clinical use or human consumption