Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetate Buffer, for Aluminum (Eriochrome Cyanine R Method), Ricca Chemical
SDP

Catalog No. 5916 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5916 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 5916 Supplier Ricca Chemical Company Supplier No. 5916
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Acetate buffer solution formulated for use in the Eriochrome Cyanine R method for aluminum determination.

Specifications

Chemical Name or Material Acetate Buffer
Name Note for Aluminum Determination
CAS 6131-90-4, 64-19-7, 7732-18-5
Color Colorless
Concentration 1X
For Use With (Application) Aluminum Analysis
pH 4-6
Physical Form Liquid
Quantity 500 mL
Solution Type Buffer
Packaging Natural Poly Bottle
Grade Laboratory
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.