Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetate Buffer, pH 3.5 For USP Heavy Metals Test, Reagents

Catalog No. p-200021766
Change view
Click to view available options
Quantity:
120 mL
500 mL
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-335-948 120 mL
01-335-944 4 L
01-335-943 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-335-948 Supplier Reagents Holdings Llc Supplier No. CA102300120A
Only null left
Add to Cart
Add to Cart

Acetate Buffer, pH 3.5 For USP Heavy Metals Test, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Acetate Buffer, pH 3.5 For USP Heavy Metals Test
CAS 7732-18-5, 631-61-8, 631-61-8
Boiling Point 118°C
Color Colorless
pH 3.5
Physical Form Liquid
Quantity 120 mL
Solution Type Buffer
Packaging Poly Bottle
Specific Gravity 1.0737
Sterility Non-sterile
Grade Reagent
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.