Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetate Buffer, pH 4.6, for Bromide Analysis, Ricca Chemical
SDP

Catalog No. 5432 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
54-32 1 L
5416 500 mL
541 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 54-32 Supplier Ricca Chemical Company Supplier No. 5432
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For Analysis of Bromide

Specifications

Chemical Name or Material Acetate Buffer
Name Note pH 4.6
CAS 7732-18-5, 7647-14-5, 6131-90-4, 64-19-7
CAS Min % 82.0, 7.42, 5.61, 2.59
Color Colorless
CAS Max % 86.0, 7.72, 5.83, 2.7
Concentration 1X
For Use With (Application) Water Analysis
pH 4.6
Physical Form Liquid
Quantity 1 L
Solution Type Buffer
Packaging Natural Poly Bottle
Grade Laboratory
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.