Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetate Buffer Solution, pH 3.5, for USP Heavy Metals Testing, Ricca Chemical
SDP

Catalog No. R0060100250 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
250 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R0060100250 250 mL
R0060100120 120 mL
60132 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. R0060100250 Supplier Ricca Chemical Company Supplier No. R0060100250A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Chemical Name or Material Acetate Buffer Solution
CAS 7732-18-5, 631-61-8, 7647-01-0
CAS Max % 65.8, 23.81, 10.4
Color Colorless Liquid
Concentration 1X
For Use With (Application) Water Analysis
pH 3.5
Physical Form Liquid
Quantity 250 mL
Solution Type Buffer
Packaging Natural Poly Bottle
Sterility Non-sterile
Grade Laboratory
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.