Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetate Buffer Solution, pH 3.5, For USP Heavy Metals Testing, Ricca Chemical

Click to view available options
Quantity:
1L
Packaging:
Poly natural
126-96-5,7647-01-0,631-61-8
C4H7NaO4
142.09
BHZOKUMUHVTPBX-UHFFFAOYSA-M
87315833
126-96-5,7647-01-0,631-61-8
C4H7NaO4
142.09
BHZOKUMUHVTPBX-UHFFFAOYSA-M
87315833
CAS Min % 63.0,18.71,15.87
CAS Max % 66.0,19.47,16.51
Physical Form Liquid
Packaging Poly natural
Color Colorless
Quantity 1L
Chemical Name or Material Acetate Buffer Solution

ShelfLife : 24 Months