Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetate Buffer Solution pH 4.0, Lovibond™

Catalog No. 09534705 Shop All Tintometer Group Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-534-705 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-534-705 Supplier Tintometer Group Supplier No. R50002
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Acetate Buffer Solution
CAS 64-19-7, 127-09-3, 7732-18-5
Color Colorless
Physical Form Liquid
Quantity 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.