Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACETATE BUFFER SOLUTION PH 4.65, SODIUM ACETATE / ACETIC ACID, Solstice

Catalog No. 60045900 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
6 x 1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-045-900 6 x 1 L
60-022-96 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-045-900 Supplier Solstice Supplier No. 311036X1L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SODIUM ACETATE / ACETIC ACID

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material ACETATE BUFFER SOLUTION PH 4.65
Name Note SODIUM ACETATE / ACETIC ACID
CAS 127-09-3, 64-19-7, 7732-18-5
CAS Min % ≥90.0000%
CAS Max % ≤100.0000%, <1.0000%
Density 1 g/cm3
Quantity 6 x 1 L
Solution Type Buffer
Linear Formula HNO3
Packaging Poly Bottle
MDL Number MFCD00011349
Beilstein 3786729
Synonym Sodium acetate buffer solution, Michaelis buffer
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.