Learn More
Apexbio Technology LLC Acetate gossypol 12542-36-8 100mg
SDP
Catalog No. 5000094598
Encompass_Preferred
Acetate gossypol (CAS 12542-36-8) is a polyphenolic compound isolated from the seeds and bark of Gossypium herbaceum L It functions primarily as an inhibitor of anti-apoptotic Bcl-2 family proteins thereby promoting apoptosis in various cancer cell types By disrupting mitochondrial membrane potential and modulating cell survival pathways acetate gossypol has been utilized in studies investigating mechanisms of programmed cell death and antitumor activity This compound is frequently employed as a molecular tool in cancer biology and apoptosis research Store under sealed cool and dry conditions to ensure stability
Catalog No. 50-000-94598 Supplier Apexbio Technology LLC Supplier No. N2134100
Acetate gossypol (CAS 12542-36-8) is a polyphenolic compound isolated from the seeds and bark of Gossypium herbaceum L It functions primarily as an inhibitor of anti-apoptotic Bcl-2 family proteins thereby promoting apoptosis in various cancer cell types By disrupting mitochondrial membrane potential and modulating cell survival pathways acetate gossypol has been utilized in studies investigating mechanisms of programmed cell death and antitumor activity This compound is frequently employed as a molecular tool in cancer biology and apoptosis research Store under sealed cool and dry conditions to ensure stability