Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Acetate kinase (ACK) | 9027-42-3 | 45 kDa | 200 U
SDP

Catalog No. 5000398864
Encompass_Preferred

Acetate kinase (ACK) is an enzyme widely distributed in bacteria and archaea. It catalyzes the phosphorylation of acetate and promotes the production of acetyl-CoA in the presence of ATP and divalent cations. This enzyme is often used in biochemical research.

  • Expressed by recombinant Escherichia coli
  • Features an N-terminal His-tag
  • Molecular weight of 45 kDa
  • Buffer components: 20 mM Tris-HCl, pH 7.5, 10% glycerol
  • Catalyzes acetic acid to produce acetyl phosphate

Catalog No. 50-003-98864 Supplier Medchemexpress LLC Supplier No. HYP2887200U
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Acetate kinase (ACK) is an enzyme widely distributed in bacteria and archaea. It catalyzes the phosphorylation of acetate and promotes the production of acetyl-CoA in the presence of ATP and divalent cations. This enzyme is often used in biochemical research.

  • Expressed by recombinant Escherichia coli
  • Features an N-terminal His-tag
  • Molecular weight of 45 kDa
  • Buffer components: 20 mM Tris-HCl, pH 7.5, 10% glycerol
  • Catalyzes acetic acid to produce acetyl phosphate

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.