Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Acetate kinase (ACK) | 9027-42-3 | 45 kDa | 50 U
SDP

Catalog No. 5000399456
Encompass_Preferred

Acetate kinase (ACK) is an enzyme widely distributed in bacteria and archaea. In the presence of ATP and divalent cations, it catalyzes the phosphorylation of acetate and promotes the production of acetyl-CoA. This product, expressed by recombinant Escherichia coli, is often used in biochemical research.

  • Catalyzes phosphorylation of acetate
  • Promotes the production of acetyl-CoA
  • Expressed by recombinant Escherichia coli with an N-terminal His-tag
  • Molecular weight of 45 kDa
  • Buffer components: 20 mM Tris-HCl, pH 7.5, 10% glycerol

Catalog No. 50-003-99456 Supplier Medchemexpress LLC Supplier No. HYP288750U
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Acetate kinase (ACK) is an enzyme widely distributed in bacteria and archaea. In the presence of ATP and divalent cations, it catalyzes the phosphorylation of acetate and promotes the production of acetyl-CoA. This product, expressed by recombinant Escherichia coli, is often used in biochemical research.

  • Catalyzes phosphorylation of acetate
  • Promotes the production of acetyl-CoA
  • Expressed by recombinant Escherichia coli with an N-terminal His-tag
  • Molecular weight of 45 kDa
  • Buffer components: 20 mM Tris-HCl, pH 7.5, 10% glycerol

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.