Learn More
Sigma Aldrich Fine Chemicals Biosciences Acetate Kinase from Escher
Supplier:  Sigma Aldrich Fine Chemicals Biosciences A74371KU
Encompass
Acetate kinase belongs to acetate and sugar kinase/Hsc70/actin (ASKHA) structural superfamily. It exists as a dimer and has nucleotide binding site in the cleft between N and C-terminal domain.
Catalog No. 50-290-3620