Learn More
Sigma Aldrich Fine Chemicals Biosciences Acetate Kinase from Escher
Catalog No. 502899100
Encompass
Acetate kinase belongs to acetate and sugar kinase/Hsc70/actin (ASKHA) structural superfamily. It exists as a dimer and has nucleotide binding site in the cleft between N and C-terminal domain.
Catalog No. 50-289-9100 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. A7437250UN
Acetate kinase belongs to acetate and sugar kinase/Hsc70/actin (ASKHA) structural superfamily. It exists as a dimer and has nucleotide binding site in the cleft between N and C-terminal domain.