Learn More
Selleck Chemical LLC Acetazolamide S4506-200mg
SDP
Catalog No. 503108133
Encompass_Preferred
Acetazolamide (Diamox) a potent carbonic anhydrase (CA) inhibitor is commonly used in clinical practice as an immediate and readily available option for acute reduction of intraocular pressure(IOP)
Catalog No. 50-310-8133 Supplier Selleck Chemical LLC Supplier No. S4506200MG
Acetazolamide (Diamox) a potent carbonic anhydrase (CA) inhibitor is commonly used in clinical practice as an immediate and readily available option for acute reduction of intraocular pressure(IOP)