Learn More
Selleck Chemical LLC Acetazolamide S4506-1g
SDP
Catalog No. 503112189
Encompass_Preferred
Acetazolamide (Diamox) a potent carbonic anhydrase (CA) inhibitor is commonly used in clinical practice as an immediate and readily available option for acute reduction of intraocular pressure(IOP)
Catalog No. 50-311-2189 Supplier Selleck Chemical LLC Supplier No. S45061G
Acetazolamide (Diamox) a potent carbonic anhydrase (CA) inhibitor is commonly used in clinical practice as an immediate and readily available option for acute reduction of intraocular pressure(IOP)