Learn More
Medchemexpress LLC Acetazolamide | 59-66-5 | 99.2% | 1 ML
SDP
Catalog No. 5000408891
Encompass_Preferred
Acetazolamide is a carbonic anhydrase (CA) IX inhibitor with an IC50 of 30 nM for hCA IX. It exhibits diuretic, antihypertensive, and anti-gonococcal activities, and also inhibits hCA II with an IC50 of 130 nM.
  • Carbonic anhydrase (CA) IX inhibitor
  • IC50 of 30 nM for hCA IX
  • Possesses diuretic, antihypertensive, and anti-gonococcal activities
  • Also inhibits hCA II with an IC50 of 130 nM
  • Small heteroaromatic sulfonamide
  • Binds to various carbonic anhydrases with high affinity
  • For research use only
Catalog No. 50-004-08891 Supplier Medchemexpress LLC Supplier No. HYB078210MM/1ML
Acetazolamide is a carbonic anhydrase (CA) IX inhibitor with an IC50 of 30 nM for hCA IX. It exhibits diuretic, antihypertensive, and anti-gonococcal activities, and also inhibits hCA II with an IC50 of 130 nM.
  • Carbonic anhydrase (CA) IX inhibitor
  • IC50 of 30 nM for hCA IX
  • Possesses diuretic, antihypertensive, and anti-gonococcal activities
  • Also inhibits hCA II with an IC50 of 130 nM
  • Small heteroaromatic sulfonamide
  • Binds to various carbonic anhydrases with high affinity
  • For research use only