Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Acetazolamide | 59-66-5 | 99.7% | 222.25 | C4H6N4O3S2 | 10 MG
SDP

Catalog No. 5000297429
Encompass_Preferred

Acetazolamide is supplied as an analytical reference standard intended for research and analytical applications, including assay development, method validation, and quality control. The material is provided in a 10 mg vial and is suitable for chromatographic and mass spectrometry workflows.

  • Analytical reference standard suitable for HPLC, GC, and MS.
  • High reported purity (99.69%), supporting accurate quantitation.
  • Small 10 mg vial format for calibration and trace analysis.
  • Molecular formula C4H6N4O3S2 and molecular weight 222.25 g/mol.
  • CAS number 59-66-5 for unambiguous substance identification.

Catalog No. 50-002-97429 Supplier Medchemexpress LLC Supplier No. HYB0782R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Acetazolamide is supplied as an analytical reference standard intended for research and analytical applications, including assay development, method validation, and quality control. The material is provided in a 10 mg vial and is suitable for chromatographic and mass spectrometry workflows.

  • Analytical reference standard suitable for HPLC, GC, and MS.
  • High reported purity (99.69%), supporting accurate quantitation.
  • Small 10 mg vial format for calibration and trace analysis.
  • Molecular formula C4H6N4O3S2 and molecular weight 222.25 g/mol.
  • CAS number 59-66-5 for unambiguous substance identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.