Learn More
Medchemexpress LLC Acetazolamide | 59-66-5 | 99.9% | 500 MG
SDP
Catalog No. 5000408250
Encompass_Preferred
Acetazolamide is a carbonic anhydrase (CA) IX inhibitor with an IC50 of 30 nM for hCA IX. It exhibits diuretic, antihypertensive, and anti-gonococcal activities. This small heteroaromatic sulfonamide binds to various carbonic anhydrases with high affinity. It is intended for research use only.
  • Inhibits carbonic anhydrase (CA) IX and hCA II.
  • Exhibits diuretic properties.
  • Demonstrates antihypertensive activity.
  • Possesses anti-gonococcal activities.
  • Suitable for various research applications.
Catalog No. 50-004-08250 Supplier Medchemexpress LLC Supplier No. HYB0782500MG
Acetazolamide is a carbonic anhydrase (CA) IX inhibitor with an IC50 of 30 nM for hCA IX. It exhibits diuretic, antihypertensive, and anti-gonococcal activities. This small heteroaromatic sulfonamide binds to various carbonic anhydrases with high affinity. It is intended for research use only.
  • Inhibits carbonic anhydrase (CA) IX and hCA II.
  • Exhibits diuretic properties.
  • Demonstrates antihypertensive activity.
  • Possesses anti-gonococcal activities.
  • Suitable for various research applications.