Learn More
Abcam Acetazolamide, carbonic anhydrase inhibitor, 5MG
Catalog No. 502676163
Encompass_Preferred
MW 222.3 Da, Purity >98%. Non-specific carbonic anhydrase inhibitor with diverse physiological effects that are cell and tissue specific. Heterocyclic primary sulfonamide.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-6163 Supplier Abcam Supplier No. AB2301495MG
MW 222.3 Da, Purity >98%. Non-specific carbonic anhydrase inhibitor with diverse physiological effects that are cell and tissue specific. Heterocyclic primary sulfonamide.
The product is subject to the following: Abcam Restricted Use Statement