Learn More
Medchemexpress LLC Acetazolamide-d3 | 1189904-01-5 | 99.9% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB0782S1MG
Encompass_Preferred
Acetazolamide-d3 is a deuterium-labeled form of Acetazolamide, a carbonic anhydrase (CA) IX inhibitor with an IC50 of 30 nM for hCA IX. It exhibits diuretic, antihypertensive, and anti-gonococcal activities. The incorporation of stable heavy isotopes, like deuterium, into drug molecules is primarily for quantitation during the drug development process, with the potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Suitable for use as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Possesses diuretic activity
  • Possesses antihypertensive activity
  • Possesses anti-gonococcal activities
Catalog No. 50-004-09278