Learn More
Cayman Chemical AcetazolamIde-d3 1mg
SDP
Catalog No. 502944055
Encompass_Preferred
An internal standard for the quantification of acetazolamide by GC- or LC-MS
Catalog No. 50-294-4055 Supplier Cayman Chemical Supplier No. 401251 MG
An internal standard for the quantification of acetazolamide by GC- or LC-MS